Learn More
p-Toluenesulfinic acid sodium salt, 97% (dry wt.), water <5%
Description
p-Toluenesulfinic acid sodium salt is used as an intermediate for the preparation of pharmaceuticals and dispersal dyes. It acts as a curing agent of grouting material. It is also used as a discharge medium of dyes and pigments. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
p-Toluenesulfinic acid sodium salt is used as an intermediate for the preparation of pharmaceuticals and dispersal dyes. It acts as a curing agent of grouting material. It is also used as a discharge medium of dyes and pigments. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 25 g |
| Beilstein | 4621454 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 178.19 |
| Percent Purity | 97% |
| Assay | (dry wt.) |
| Chemical Name or Material | p-Toluenesulfinic acid sodium salt, Water <5% |
Safety and Handling
| EINECSNumber | 212-538-5 |
| RTECSNumber | XT4725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.