Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonamide 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
70-55-3
C7H9NO2S
171.21
MFCD00011692
LMYRWZFENFIFIT-UHFFFAOYSA-N
p-toluenesulfonamide,4-toluenesulfonamide,tosylamide,p-tosylamide,benzenesulfonamide, 4-methyl,p-tolylsulfonamide,toluene-4-sulfonamide,tolylsulfonamide,4-methylbenzene-1-sulfonamide,p-toluenesulfamide
6269
CHEBI:34435
4-methylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)N
70-55-3
C7H9NO2S
171.21
MFCD00011692
LMYRWZFENFIFIT-UHFFFAOYSA-N
p-toluenesulfonamide,4-toluenesulfonamide,tosylamide,p-tosylamide,benzenesulfonamide, 4-methyl,p-tolylsulfonamide,toluene-4-sulfonamide,tolylsulfonamide,4-methylbenzene-1-sulfonamide,p-toluenesulfamide
6269
CHEBI:34435
4-methylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)N
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 202°C
Color White
Quantity 250g
Melting Point 136.0°C to 140.0°C
Boiling Point 221.0°C (10.0 mmHg)
Assay Percent Range 99%
Linear Formula CH3C6H4SO2NH2
Beilstein 11, 104
Fieser 13,294
Solubility Information Solubility in water: 0.32 g/100 mL(25° c). Other solubilities: soluble in alcohol
Formula Weight 171.21
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material p-Toluenesulfonamide
Show More Show Less

GHS H Statement
Harmful in contact with skin. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation. Harmful if inhaled. Contact with acids liberates very toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning