Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic acid, sodium salt

Catalog No. p-3213115
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-84-1
Molecular Formula C7H7NaO3S
Molecular Weight (g/mol) 194.18
MDL Number MFCD00798566,MFCD00064388
InChI Key KVCGISUBCHHTDD-UHFFFAOYSA-M
Synonym sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34
PubChem CID 3720192
IUPAC Name sodium;4-methylbenzenesulfonate
SMILES [Na+].CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >400°C
Color White
Flash Point >500°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 11,97
Fieser 11,535; 12,507
Merck Index 14,9533
Solubility Information Solubility in water: soluble.
Formula Weight 194.19
Percent Purity ≥97.5% (ion exchange; alkalimetry)
Physical Form Powder
Chemical Name or Material p-Toluenesulfonic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 211-522-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.