Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoroacetic acid, 99+%, extra pure
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -15.0°C |
Boiling Point | 72.0°C |
Color | Colorless |
Assay Percent Range | 0.99 |
Linear Formula | CF3CO2H |
Beilstein | 02, II, 186 |
Fieser | 01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340 |
Merck Index | 15, 9847 |
Solubility Information | Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane |
IUPAC Name | trifluoroacetic acid |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if inhaled. Harmful to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
EINECSNumber | 200-929-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction