Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonyl Isocyanate 95.0+%, TCI America™
SDP

Catalog No. T0998100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0998100G 100 g
T099825G 25 g
T0998500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0998100G Supplier TCI America Supplier No. T0998100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4083-64-1
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00002030
InChI Key VLJQDHDVZJXNQL-UHFFFAOYSA-N
Synonym tosyl isocyanate, p-toluenesulfonyl isocyanate, 4-methylbenzenesulfonyl isocyanate, p-toluenesulphonyl isocyanate, benzenesulfonyl isocyanate, 4-methyl, tosylisocyanate, p-tolylsulfonyl isocyanate, unii-h9004fjx6v, p-toluenesulfonic acid, anhydride with isocyanic acid, p-toluene sulfonyl isocyanate
PubChem CID 77703
IUPAC Name 4-methylbenzene-1-sulfonyl isocyanate
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 130°C
Quantity 100 g
Formula Weight 197.21
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material p-Toluenesulfonyl Isocyanate
EINECSNumber (3)-2222
RTECSNumber DB9032000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.