Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenethiol, 98%

Catalog No. AC138710025
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Pail

CAS: 106-45-6 | C7H8S | 124.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-45-6
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.2
MDL Number MFCD00004851
InChI Key WLHCBQAPPJAULW-UHFFFAOYSA-N
Synonym p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol
PubChem CID 7811
IUPAC Name 4-methylbenzenethiol
SMILES CC1=CC=C(C=C1)S

Specifications

Melting Point 40.0°C to 44.0°C
Color White to Yellow
Boiling Point 195.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic Pail
Linear Formula CH3C6H4SH
Quantity 2.5 kg
Beilstein 06, 416
Merck Index 15, 9478
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 124.2
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material p-Toluenethiol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air
EINECSNumber 203-399-1
RTECSNumber XT8925000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.