Alfa Aesar™ p-Tolunitrile, 98+%

Manufacturer: Alfa Aesar™ B2302918

Quantity 50g

 View more versions of this product

Catalog No. AAB2302918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104-85-8
    Molecular Formula C8H7N
    Formula Weight 117.15
    MDL Number MFCD00001827
    Beilstein 507386
    Merck Index 14,9538
    Synonym 4-Cyanotoluene; 4-Methylbenzonitrile
    Chemical Name or Material p-Tolunitrile

    Specifications

    Formula Weight 117.15
    MDL Number MFCD00001827
    Synonym 4-Cyanotoluene; 4-Methylbenzonitrile
    Boiling Point 217°C
    Melting Point 26° to 29°C
    Quantity 50g
    TSCA TSCA
    Molecular Formula C8H7N
    CAS 104-85-8
    Beilstein 507386
    Merck Index 14,9538
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number XV0700000
    Flash Point 85°C (185°F)
    Odor Almond-like
    Chemical Name or Material p-Tolunitrile
    Assay ≥98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit