Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p-Tolyl isocyanate, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Brown |
Density | 1.0560g/mL |
Boiling Point | 70.0°C to 72.0°C (10.0 mmHg) |
Flash Point | 66°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Refractive Index | 1.5301 to 1.5321 |
Linear Formula | CH3C6H4NCO |
Quantity | 25 g |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Fatal if inhaled. May cause respiratory irritation. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Very toxic |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water |
EINECSNumber | 210-743-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction