Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl isocyanate, 99%

Catalog No. p-4510390
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 622-58-2 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-58-2
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key MGYGFNQQGAQEON-UHFFFAOYSA-N
Synonym p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzene
PubChem CID 69325
ChEBI CHEBI:53732
IUPAC Name 1-isocyanato-4-methylbenzene
SMILES CC1=CC=C(C=C1)N=C=O

Specifications

Color Brown
Density 1.0560g/mL
Boiling Point 70.0°C to 72.0°C (10.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5301 to 1.5321
Linear Formula CH3C6H4NCO
Quantity 5 g
Beilstein 12,955
Specific Gravity 1.056
Formula Weight 133.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material p-Tolyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very toxic
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 210-743-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.