Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoroacetic acid, 99+%, for spectroscopy
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Specifications
| Absorbance | 0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm |
| Melting Point | -15.0°C |
| Boiling Point | 72.0°C |
| Color | Colorless |
| Linear Formula | CF3CO2H |
| Beilstein | 02,IV,458 |
| Fieser | 10,418; 01,1219; 17,340; 16,83; 15,338; 14,322; 12,529; 11,557; 09,483; 08,503; 07,388; 06,613; 05,695; 04,530; 02,433; 03,305 |
| Merck Index | 159847 |
| Solubility Information | Solubility in water: miscible. Other solubilities: miscible with ether,acetone,ethanol,benzene,,ccl4,hexane,methanol,xylene and hexane |
| Formula Weight | 114.02 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if inhaled. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
| EINECSNumber | 200-929-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction