Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene-2-sulfonic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. D0681500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0681500G 500 g
D068125G 25 g
D0681100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0681500G Supplier TCI America Supplier No. D0681500G

Chemical Identifiers

CAS 609-54-1
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00151010
InChI Key IRLYGRLEBKCYPY-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641
PubChem CID 11868
IUPAC Name 2,5-dimethylbenzene-1-sulfonic acid
SMILES CC1=CC=C(C)C(=C1)S(O)(=O)=O

Specifications

Melting Point 86°C
Color Red-White
Quantity 500 g
Physical Form Crystal-Powder at 20°C
Formula Weight 186.23
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material p-Xylene-2-sulfonic Acid Hydrate
EINECSNumber (9)-2093
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.