Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene, 99%

Catalog No. aaa10534ae
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 106-42-3 | C8H10 | 106.17 g/mol

p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.

Solubility
Miscible with alcohol, ether, acetone, benzene and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 12°C to 13°C
Density 0.866
Boiling Point 137°C to 138°C
Quantity 100 mL
Flash Point 27°C (80°F)
Odor Aromatic
Refractive Index 1.495
UN Number UN1307
Beilstein 1901563
Merck Index 14,10081
Solubility Information Miscible with alcohol,ether,acetone,benzene and chloroform. Immiscible with water.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material p-Xylene
Show More Show Less
Hazard Category H226-H304-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
EINECSNumber 203-396-5
RTECSNumber ZE2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.