Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene, 99%, for HPLC

Catalog No. ac445681000
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle

CAS: 106-42-3 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 13.0°C
Absorbance 0.01 max. at 380nm, 0.01 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 320nm, 1 max. at 294nm
Density 0.8660g/mL
Boiling Point 138.0°C
Quantity 100 mL
Flash Point 25°C
Packaging Glass Bottle
Refractive Index 1.4940 to 1.4960
Assay Percent Range 99%
Specific Gravity 0.866
Linear Formula C6H4(CH3)2
Viscosity 0.648 mPa.s (20°C)
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Percent Purity 99%
Grade HPLC
Chemical Name or Material p-Xylene
Show More Show Less
Hazard Category Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.