Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
GSA_VA

Catalog No. BP327500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP327-500 500 g Poly Bottle
BP327-1 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP327-500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6132-04-3 | C6H9Na3O9 | 294.10 g/mol

Sodium Citrate Dihydrate (Small, Colorless Granules)

Sodium Citrate is used primarily in the preparation of SSC buffer used in Southern transfers.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-04-3 , 68-04-2
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Decomposition Information >230°C
Melting Point 300°C
Color White
pH 7.5 to 9.5
Assay Percent Range 99.0 to 101.0 %
Packaging Poly Bottle
Quantity 500 g
Merck Index 15, 8737
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 294.09
Percent Purity 99.0 to 101.0%
Physical Form Solid
Chemical Name or Material Sodium Citrate Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.