Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes (Certified ACS), Fisher Chemical™

Catalog No. X51 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
1 gal.
4 L
20 L
50 L
200 L
Packaging:
BasicPak
Glass Bottle
Poly Bottle
Safe-Cote™ Amber Glass 
Steel Drum
Steel Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
X5-1 1 L Glass Bottle
X5-4 4 L Glass Bottle
X5-20 20 L Steel Pail
X5FB50 50 L BasicPak
X5-200 200 L Steel Drum
X5-500 500 mL Glass Bottle
X5SK-4 4 L Safe-Cote™ Amber Glass 
X5P-1GAL 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. X5-1
Only null left
Add to Cart

Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
MDL Number MFCD00077264

Specifications

Melting Point -34°C
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range Assay: ≥98.5%
Residue after Evaporation 0.002% max.
Molecular Formula C8H10
Vapor Pressure 8.29mmHg at 25°C
Water 0.05% max.
Synonym Xylol, Dimethylbenzene
Boiling Point 136°C
Formula Weight 106.17
Percent Purity ≥98.5%
Grade Assay: ≥98.5%
Chemical Name or Material Xylenes
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.