Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p107 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$336.16 - $690.48
Specifications
| Antigen | p107 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
p107 Polyclonal specifically detects p107 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| p107 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| cellular protein 107,107 kDa retinoblastoma-associated protein, cp107, p107MGC40006, PRB1, retinoblastoma-like 1 (p107), retinoblastoma-like protein 1 | |
| RBL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P28749 | |
| 5933 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ESLAWSHDSALWEALQVSANKVPTCEEVIFPNNFETGNGGNVQGHLPLMPMSPLMHPRVKEVRTDSGSLRRDMQPLSPISVHER | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title