Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p107 Polyclonal Antibody, Huabio

Rabbit Polyclonal Antibody

Supplier:  HUABIO RESEARCH INC HA500481

Encompass
Encompass_Preferred

Catalog No. 50-254-4540


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

p107 Polyclonal antibody specifically detects p107 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

p107
Polyclonal
Unconjugated
RBL1
107 kDa retinoblastoma-associated protein; AW547426; CP107; DNA-binding protein RFX5; MGC40006; p107; pRb1; RB transcriptional corepressor like 1; Rbl1; Regulatory factor X 5; regulatory factor X, 5 (influences HLA class II expression); regulatory factor X5; retinoblastoma-like 1; retinoblastoma-like 1 (p107); retinoblastoma-like protein 1; RFX5
Recombinant protein within human p107 aa 869-1,068/1,068.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
P28749
Rabbit
Antigen affinity chromatography
RUO
5933
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.