Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p114RhoGEF (ARHGEF18) Monoclonal Antibody (OTI4B5), OriGene
SDP

Catalog No. 502544023
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4023 Supplier OriGene Supplier No. CF810293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p114RhoGEF (ARHGEF18) Monoclonal antibody specifically detects p114RhoGEF (ARHGEF18) in Human samples. It is validated for Western Blot

Specifications

Antigen p114RhoGEF (ARHGEF18)
Applications Western Blot
Classification Monoclonal
Clone OTI4B5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SLC2A4RG
Gene Accession No. Q9NR83
Gene Alias 114 kDa Rho-specific guanine nucleotide exchange factor; AI467246; Arhgef18; D030053O22Rik; KIAA0521; p114RhoGEF; P114-RhoGEF; p114-Rho-GEF; rho guanine nucleotide exchange factor 18; rho/rac guanine nucleotide exchange factor (GEF) 18; Rho/Rac guanine nucleotide exchange factor 18; Rho-specific guanine nucleotide exchange factor p114; SA-RhoGEF; Septin-associated RhoGEF
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-259 of human SLC2A4RG produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56731
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.