Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p120-catenin Antibody (25a), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254411MFV610
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
p120-catenin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
25a | |
Flow Cytometry | |
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
Mouse p120-catenin (pY96) (Uniprot: O60716) | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
1500 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction