Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p120-catenin Antibody (25a), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254411PECY55
Description
p120-catenin Monoclonal specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
p120-catenin | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CTNND1 | |
Mouse p120-catenin (pY96) (Uniprot: O60716) | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 κ |
Flow Cytometry | |
25a | |
Flow Cytometry | |
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
Mouse | |
Protein A or G purified | |
Phospho Specific, Signal Transduction | |
1500.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction