Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p120-catenin Antibody (M354), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323202
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
p120-catenin | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
M354 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:50 | |
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
Clone (M354) was generated from a peptide that includes amino acids from the central region of human p120-catenin. This peptide sequence is highly conserved in rat and mouse p120-catenin. [UniProt# O60716] | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
1500 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction