Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p120-catenin Antibody (SR1036), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321614100UL
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
p120-catenin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1036 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:30 | |
Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
A synthesized peptide derived from human p120-catenin (Uniprot #: O60716) | |
100 μg | |
Phospho Specific, Signal Transduction | |
1500 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction