Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p130, p Thr986 Antibody (SR1109), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321608100UL
Description
p130 Monoclonal antibody specifically detects p130 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
p130 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
SR1109 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
FLJ26459, P130, PRB2, RB2, RBR-2, retinoblastoma-like 2 (p130), retinoblastoma-like protein 2,130 kDa retinoblastoma-associated protein, Retinoblastoma-related protein 2 | |
A synthesized peptide derived from human phospho-p130[Thr986] (Uniprot number: Q08999) | |
100 μL | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
5934 | |
Store at -20° C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction