Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p130 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA132114
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132114 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132114 Supplier Invitrogen™ Supplier No. PA132114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as a concentrated solution. Centrifuge briefly prior to opening vial.

RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p130
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene RBL2
Gene Accession No. Q08999, Q64700
Gene Alias 130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RB transcriptional corepressor like 2; Rb2; rb2 p130; Rbl2; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130); retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Symbols RBL2
Host Species Rabbit
Immunogen Rb2 (p130) peptide corresponding to a region near the C-terminus of the human protein conjugated to KLH.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19651, 5934
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.