Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p130 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578147 Supplier Invitrogen™ Supplier No. PA578147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Jurkat, Jurkat nuclear Predicted Reactivity: Bovine (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p130
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene RBL2
Gene Accession No. O55081, Q08999, Q64700
Gene Alias 130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RB transcriptional corepressor like 2; Rb2; rb2 p130; Rbl2; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130); retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Symbols RBL2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Rb2/p130. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19651, 5934, 81758
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.