Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p130Cas Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS855R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS855R7 7 mL Unconjugated Prediluted Prediluted
MS855P0 100 μL Unconjugated Concentrated 200μg/mL
MS855P1 500 μL Unconjugated Concentrated 200μg/mL
MS855P 1 mL Unconjugated Concentrated 200μg/mL
MS855P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS855PABX 200 μL Unconjugated Concentrated 1mg/mL
MS855R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS855R7 Supplier Epredia Supplier No. MS855R7

Take advantage of a quality product that delivers accurate results with the Epredia™ p130Cas Ab-1, Mouse Monoclonal Antibody.

The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich region p130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.

Host Species: Mouse

Clone: CAS-14; same as 35B-1A4

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human p130Cas protein

Molecular Weight: 130kDa

Positive Control: MCF-7 Cells, HUVEC cells, Tonsil or breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p130superscriptCassuperscript Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CAS-14; same as 35B-1A4
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human p130superscriptCassuperscript protein
Purification Method MCF-7 cells, HUVEC cells, tonsil or breast CA
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.