Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p130Cas/BCAR1 Antibody (GT331), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313565
This item is not returnable.
View return policy
Description
p130Cas/BCAR1 Monoclonal antibody specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| p130Cas/BCAR1 | |
| Monoclonal | |
| Unconjugated | |
| breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
| Recombinant protein encompassing a sequence within the center region of human p130Cas/BCAR1. The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| GT331 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 9564 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction