Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p130Cas/BCAR1 Antibody (M144), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323253
Description
p130Cas/BCAR1 Monoclonal antibody specifically detects p130Cas/BCAR1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
p130Cas/BCAR1 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
M144 | |
Western Blot 1:1000, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:100 | |
breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
Clone M144 was generated from a recombinant protein containing amino acid residues in the C-terminal region of rat p130Cas/BCAR1. This rat sequence is highly conserved in human, mouse, and chicken p130Cas/BCAR1. [UniProt# Q63767] | |
0.1 mL | |
Phospho Specific | |
9564 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction