Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p14ARF/CDKN2A Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059085G
Description
p14ARF/CDKN2A Polyclonal specifically detects p14ARF/CDKN2A in Human samples. It is validated for Western Blot.Specifications
p14ARF/CDKN2A | |
Polyclonal | |
Western Blot | |
Q8N726 | |
CDKN2A | |
A synthetic peptide made to a portion of human p14ARF (between residues 100-173). [Swiss-Prot# Q8N726] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, P16, p16-INK4, p16-INK4a, P16INK4A, P19, p19Arf, TP16 | |
Rabbit | |
16 kDa | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
1029 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction