Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p14ARF/CDKN2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | p14ARF/CDKN2A |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2500, ELISA 1:100-1:2000, Immunohistochemistry 1:500-1:1000, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:500-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Description
p14ARF/CDKN2A Polyclonal specifically detects p14ARF/CDKN2A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
p14ARF/CDKN2A | |
Western Blot 1:500-1:2500, ELISA 1:100-1:2000, Immunohistochemistry 1:500-1:1000, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
NP_478102.1 | |
1029 | |
The epitope recognized by this antibody maps to a region between residue 25 and 75 of human p14ARF using the numbering given in entry Uniprot ID Q8N726. | |
Primary | |
Store at 4C. Do not freeze. |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, P16, p16-INK4, p16-INK4a, P16INK4A, P19, p19Arf, TP16 | |
CDKN2A | |
IgG | |
Affinity Purified | |
14 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title