Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p14ARF Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571660
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1660 10 μL
50-157-1659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1660 Supplier Bethyl Laboratories, Inc Supplier No. IHC00196T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan and White-tufted-ear marmoset

The INK4a-ARF locus encodes two unrelated proteins both of which function in tumor suppression. p14ARF arrests the cell cycle in a p53-dependent manner. p14ARF binds to mdm2 and promotes the rapid degradation of mdm2 proteinrequired for mdm2 modification and concurrent p53 stabilization and accumulation. This interaction is mediated by the exon 1Beta-encoded N-terminal domain of p14ARF and a C-terminal region of mdm2. Deletion of the ARF-INK4a locus simultaneously impairs both the INK4a-cyclin D/cdk4-RB and the ARF-mdm2-p53 pathways.

Specifications

Antigen p14ARF
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CDKN2A
Gene Accession No. Q8N726
Gene Alias ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf
Gene Symbols CDKN2A
Host Species Rabbit
Immunogen Between 125 and C-term
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.