Learn More
p14ARF Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300342AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The INK4a-ARF locus encodes two unrelated proteins both of which function in tumor suppression. p14ARF arrests the cell cycle in a p53-dependent manner. p14ARF binds to mdm2 and promotes the rapid degradation of mdm2 proteinrequired for mdm2 modification and concurrent p53 stabilization and accumulation. This interaction is mediated by the exon 1Beta-encoded N-terminal domain of p14ARF and a C-terminal region of mdm2. Deletion of the ARF-INK4a locus simultaneously impairs both the INK4a-cyclin D/cdk4-RB and the ARF-mdm2-p53 pathways.Specifications
| p14ARF | |
| Polyclonal | |
| Unconjugated | |
| CDKN2A | |
| ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1029 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8N726 | |
| CDKN2A | |
| Between 125 and C-term | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.