Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p150glued Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | p150glued |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4187
![]() |
Bethyl Laboratories, Inc
A303072AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Dynactin, a multisubunit complex, is a cytoplasmic dynein-interacting protein that functions as the 'receptor' for the dynein microtubule motor. Dynactin-dynein binding may be required for most, if not all, cytoplasmic dynein driven activities and is thought to contribute to dynein's functional diversity. Dynactin, enriched in neurons, also binds to microtubules and has been shown to function in diverse processes, including organelle transport, formation of the mitotic spindle and cytokinesis. Dynactin subunits include p22, p50, p62, p150, (also designated Glued), and Arp1.Specifications
p150glued | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DCTN1; dynactin 1 (p150, glued homolog, Drosophila); P135; DP-150; DAP-150; p150-glued; p150, Glued (Drosophila) homolog; 150 kDa dynein-associated polypeptide; dynactin 1 (p150, Glued (Drosophila) homolog); HMN7B; OTTHUMP00000202491; dynactin 1; p150, Glued homolog | |
Between 1228 and 1278 | |
Primary | |
4° C | |
DCTN1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O08788, Q14203 | |
13191, 1639 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title