Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p150glued Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501564185
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4185 Supplier Bethyl Laboratories, Inc Supplier No. A303072A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Dynactin, a multisubunit complex, is a cytoplasmic dynein-interacting protein that functions as the ′receptor′ for the dynein microtubule motor. Dynactin-dynein binding may be required for most, if not all, cytoplasmic dynein driven activities and is thought to contribute to dynein′s functional diversity. Dynactin, enriched in neurons, also binds to microtubules and has been shown to function in diverse processes, including organelle transport, formation of the mitotic spindle and cytokinesis. Dynactin subunits include p22, p50, p62, p150, (also designated Glued), and Arp1.

Specifications

Antigen p150glued
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DCTN1
Gene Accession No. O08788, Q14203
Gene Alias DCTN1; dynactin 1 (p150, glued homolog, Drosophila); P135; DP-150; DAP-150; p150-glued; p150, Glued (Drosophila) homolog; 150 kDa dynein-associated polypeptide; dynactin 1 (p150, Glued (Drosophila) homolog); HMN7B; OTTHUMP00000202491; dynactin 1; p150, Glued homolog
Gene Symbols DCTN1
Host Species Rabbit
Immunogen Between 1228 and 1278
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13191, 1639
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.