Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p15INK4b Polyclonal Antibody, Abbomax

Catalog No. 502204501
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4501 Supplier Abbomax Inc Supplier No. 620540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: 3T3, Hela Cellular location: Nucleus.

p15INK4b Polyclonal antibody specifically detects p15INK4b in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen p15INK4b
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene CDKN2B
Gene Accession No. P42772, P55271, P55272
Gene Alias AV083695; CDK inhibitory protein; CDK4B inhibitor; CDK4I; Cdkn2b; cyclin dependent kinase inhibitor 2B; Cyclin dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase 4 inhibitor B; cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase inhibitor p15; cyclin-dependent kinase inhibitor p15INK4b; cyclin-dependent kinase inhibitor protein; cyclin-dependent kinases 4 and 6 binding protein; Ink4; INK4B; MTS2; MTS-2; Multiple tumor suppressor 2; p14_CDK inhibitor; p14_INK4B; p14-INK4b; P15; p15 CDK inhibitor; p15(INK4b); p15_INK4B; p15INK4b; p15-INK4b; TP15
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of Mouse p15INK4b
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1030, 12579, 25164
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.