Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P15RS Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734105 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4105 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4105 Supplier Proteintech Group Inc Supplier No. 236521AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P15RS is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15 and may have a role in cell cycle regulation (Liu et al., 2002 ).

Specifications

Antigen P15RS
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPRD1A
Gene Accession No. Q8VDS4, Q96P16
Gene Alias HsT3101, p15INK4B related protein, P15RS, RPRD1A
Gene Symbols RPRD1A
Host Species Rabbit
Immunogen P15RS Fusion Protein Ag20409
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225283, 55197
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.