Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P16-INK4A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 50553876 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-553-876 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-553-876 Supplier Proteintech Group Inc Supplier No. 108831AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

10883-1-AP reacts with p16 specifically. P16 plays an essential role in regulating the cell cycle, and mutations in p16 increase the risk of developing various cancers, including melanoma.

Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.

Specifications

Antigen P16-INK4A
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CDKN2A
Gene Accession No. Q8N726
Gene Alias ARF, ARF,CDKN2A, CDK4 inhibitor p16 INK4, CDK4 inhibitor p16INK4, CDK4I, CDKN2, CDKN2A, CMM2, INK4, INK4a, MLM, MTS 1, MTS1, Multiple tumor suppressor 1, P16, p16 INK4, p16 INK4a, p16INK4, p16INK4a, TP16
Gene Symbols CDKN2A
Host Species Rabbit
Immunogen P16-INK4A Fusion Protein Ag1328
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.