Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p16 Mouse anti-Human, Clone: G175-405, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550834
This item is not returnable.
View return policy
Description
p16 Monoclonal antibody specifically detects p16 in Human samples. It is validated for Western blot (Routinely Tested), Immunohistochemistry-formalin (antigen retrieval required), Immunohistochemistry-frozen (Tested During Development)
Specifications
p16 | |
Monoclonal | |
31.25 μg/mL | |
Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
G175-405 | |
Unconjugated | |
p16-INK4, p16-INK4a, ARF, MTS1, CDKN2, CDK4l | |
Human p16 Recombinant Protein | |
1 mL | |
Primary | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
IgG1 |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction