Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p16INK4a/CDKN2A Mouse anti-Human, Clone: R15-A, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305757200UL
This item is not returnable.
View return policy
Description
p16INK4a/CDKN2A Monoclonal antibody specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
p16INK4a/CDKN2A | |
Monoclonal | |
Unconjugated | |
20mM Tris-HCl (pH 8.0) and 20mg/mL BSA | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
R15-A | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
Human p16INK4a/CDKN2A protein | |
200 μg | |
ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors | |
1029 | |
Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction