Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p16INK4a Monoclonal Antibody (A4A3), Huabio

Catalog No. 502544520
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4520 Supplier HUABIO RESEARCH INC Supplier No. HA600027
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p16INK4a Monoclonal antibody specifically detects p16INK4a in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen p16INK4a
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A4A3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CDKN2A
Gene Accession No. P42771
Gene Alias Alternative reading frame; Arf; ARF-INK4a; CDK4 inhibitor p16 INK4; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; Cdkn2a; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; Cyclin dependent kinase 4 inhibitor A; cyclin dependent kinase inhibitor 2A; Cyclin dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2a p16Ink4a; cyclin-dependent kinase inhibitor 2a p19Arf; cyclin-dependent kinase inhibitor 2A, isoform 1; cyclin-dependent kinase inhibitor 2A, isoform 2; cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; HGNC:1787; inhibits CDK4; INK4; INK4A; Ink4a/Arf; INK4a-ARF; isoforms 1/2/3; Melanoma p16 inhibits CDK4; mitochondrial smARF; MLM; MTS1; MTS-1; Multiple tumor suppressor 1; P14; P14 P16; P14ARF; p16; p16(INK4a); p16Cdkn2a; P16INK4; p16-INK4; p16INK4a; p16-INK4a; P19; P19 TP16; p19<ARF>; P19ARF; Pctr1; RGD:2323}; TP16; Tumor suppressor ARF
Host Species Mouse
Immunogen Recombinant protein within human p16INK4a aa 1-156.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.