Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS190P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS190P0 100 μL 200μg/mL
MS190P1 500 μL 200μg/mL
MS190P 1 mL 200μg/mL
MS190P1ABX 100 μL 1mg/mL
MS190PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS190P0 Supplier Epredia™ Supplier No. MS190P0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ensure provides accurate, reproducible results with the Epredia™ p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1, Mouse Monoclonal Antibody.

Multi-drug resistance(MDR) associated p-glycoprotein (p170) causes resistance to various drugs in a number of cancer patients. It exists in a soluble and membrane associated form. Soluble P-glycoprotein is detected in extracellular fluids of cancer patients, such as malignant ascites and serum. The Mr of soluble P-glycoprotein is the same as that of membrane bound P-glycoprotein.

Host Species: Mouse

Clone: 265/F4

Isotype: IgG1/κ

Species Reactivity: Chinese Hamster. Does not react with human. Others not known.

Epitope: Not determined

Immunogen: Colchicine-resistant viable Chinese Hamster Ovary (CHO) cells, followed by purified p170 glycoprotein from CHO cells

Molecular Weight: 170kDa

Positive Control: Drug-resistant Chinese Hamster Ovary (CHO) cells

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 265/F4
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Colchicine-resistant viable Chinese Hamster Ovary (CHO) cells, followed by purified P170 glycoprotein from CHO cells
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Hamster
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.