Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p18INK4c/CDKN2C Antibody (18P118 (DCS-118)), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1203216
Description
p18INK4c/CDKN2C Monoclonal specifically detects p18INK4c/CDKN2C in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
p18INK4c/CDKN2C | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:50, Immunofluorescence 1:10 - 1:500 | |
CDK6 inhibitor p18, CDKN6, cyclin-dependent inhibitor, cyclin-dependent kinase 4 inhibitor C, Cyclin-dependent kinase 6 inhibitor, cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4), INK4Ccyclin-dependent kinase 6 inhibitor p18, p18, p18-INK4C, p18-INK6 | |
Mouse | |
18 kDa | |
500 μL | |
Cell Cycle and Replication | |
1031 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
18P118 (DCS-118) | |
Unconjugated | |
P42773 | |
CDKN2C | |
Purified recombinant p18INK4c protein | |
Protein G purified | |
RUO | |
Primary | |
p18 (18P118 (same as DCS-118)) | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction