Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p19 INK4d Antibody (06), DyLight 550 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305845R
Description
p19 INK4d Monoclonal antibody specifically detects p19 INK4d in Human samples. It is validated for ELISASpecifications
p19 INK4d | |
Monoclonal | |
DyLight 550 | |
CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human p19 INK4d (P55273; Met 10-Leu 166). | |
0.1 mL | |
Cell Cycle and Replication, Ovarian Carcinoma Cell Markers | |
1032 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
06 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction