Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p19 INK4d Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583665 Supplier Invitrogen™ Supplier No. PA583665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLDTLKVLVE HGADVNVPDG TGALPIHLAV QEGHTAVVSF LAAESDLHRR DAR Highest antigen sequence indentity to the following orthologs: Rat - 45%, Mouse - 79%.

p19 is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well as spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p19 INK4d
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDKN2D
Gene Accession No. P55273
Gene Alias CDK inhibitor p19INK4d; CDK4I; CDKN2D; cell cycle inhibitor, Nur77 associating protein; cyclin dependent kinase inhibitor 2D; cyclin-dependent kinase 4 inhibitor D; cyclin-dependent kinase 4 inhibitor D p19; cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4); inhibitor of cyclin-dependent kinase 4d; INK4d; MTS1; p16-INK4; p16-INK4a; p19; p19INK4d; p19-INK4D; similar to cyclin-dependent kinase inhibitor 2D
Gene Symbols CDKN2D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDKN2D. Recombinant protein control fragment (Product #RP-106046).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.