Learn More
Invitrogen™ ARHGAP32 Polyclonal Antibody

Description
Immunogen sequence: ERDPSVLYQY QPHGKRQSSV TVVSQYDNLE DYHSLPQHQR GVFGGGGMGT YVPPGFPHPQ SRTYATALGQ GAFLPAELSL QHPETQIHAE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Specifications
Specifications
| Antigen | ARHGAP32 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ARHGAP32 |
| Gene Accession No. | A7KAX9 |
| Gene Alias | 3426406O18Rik; 6430596G11Rik; AI841135; ARHGAP32; Arhgap35; brain-specific Rho GTP-ase-activating protein; brain-specific Rho GTPase-activating protein; BRF2; butyrate response factor 2; EGF-response factor 2; ERF2; ERF-2; GAB-associated CDC42; GAB-associated Cdc42/Rac GTPase-activating protein; GAP-associated protein p190; Gc-gap; glucocorticoid receptor DNA binding factor 1; glucocorticoid receptor DNA-binding factor 1; Glucocorticoid receptor repression factor 1; GRF1; GRF-1; Grit; Grlf1; GTPase regulator interacting with TrkA; GTPase-activating protein for Cdc42 and Rac1; KIAA0712; KIAA1722; mKIAA0712; mKIAA1722; mRNA decay activator protein ZFP36L2; P190 RhoGAP; p190A; p190-A; p190ARHOGAP; p190RhoGAP; p200RhoGAP; p250Gap; PX-RICS; rac GTPase activating protein; rho GAP p190A; Rho GTPase activating protein 32; Rho GTPase activating protein 33 pseudogene; Rho GTPase activating protein 35; rho GTPase-activating protein 32; Rho GTPase-activating protein 35; Rho/Cdc42/Rac GTPase-activating protein RICS; rhoGAP involved in the beta-catenin-N-cadherin and NMDA receptor signaling; RhoGAP involved in the -catenin-N-cadherin and NMDA receptor signaling; rho-type GTPase-activating protein 32; RICS; RNF162C; TIS11D; TPA-induced sequence 11d; ZFP36 ring finger protein like 2; ZFP36 ring finger protein-like 2; Zfp36l2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2 |
| Gene Symbols | ARHGAP32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.