Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRF-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599556 Supplier Thermo Scientific Supplier No. PA599556
Add to Cart
Add to Cart

GRF-1 Polyclonal Antibody

GRF-1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRF-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP35
Gene Accession No. P81128, Q91YM2, Q9NRY4
Gene Alias 6430596G11Rik; AI841135; ARHGAP35; GAP-associated protein p190; glucocorticoid receptor DNA binding factor 1; Glucocorticoid receptor DNA-binding factor 1; Glucocorticoid receptor repression factor 1; GRF1; GRF-1; GRLF1; KIAA1722; LOW QUALITY PROTEIN: rho GTPase-activating protein 35; MGC10745; mKIAA1722; P190 RhoGAP; P190A; p190-A; p190ARHOGAP; p190RhoGAP; rho GAP p190A; Rho GTPase activating protein 35; rho GTPase-activating protein 35; Rho-GAP protein; p190-A; GRLF1
Gene Symbols ARHGAP35
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP35(Accession Q9NRY4), corresponding to amino acid residues Q1079-G1129.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232906, 2909, 306400
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.