Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p190RhoGEF Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566443
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6443 Supplier Bethyl Laboratories, Inc Supplier No. A303930A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a member of the Rho guanine nucleotide exchange factor family. The encoded protein interacts with low molecular weight neurofilament mRNA and may be involved in the formation of amyotrophic lateral sclerosis neurofilament aggregates. Alternate splicing results in multiple transcript variants.

Specifications

Antigen p190RhoGEF
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ARHGEF28
Gene Accession No. Q8N1W1
Gene Alias 190 kDa guanine nucleotide exchange factor, p190RhoGEF, p190-RhoGEF, RGNEF, Rho guanine nucleotide exchange factor, Rho guanine nucleotide exchange factor (GEF) 28, Rho interacting protein 2, rho-guanine nucleotide exchange factor, RIP2
Gene Symbols ARHGEF28
Host Species Rabbit
Immunogen Between 1655 and 1705
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64283
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.