Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p19ARF/CDKN2A Antibody (12-A1-1), Novus Biologicals™

Rat Monoclonal Antibody has been used in 5 publications
$206.50 - $499.50
Specifications
Antigen | p19ARF |
---|---|
Clone | 12-A1-1 |
Dilution | Western Blot 1:500, ELISA 1:5000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB200169SS
![]() |
Novus Biologicals
NB200169SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB200169
![]() |
Novus Biologicals
NB200169 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
p19ARF/CDKN2A Monoclonal specifically detects p19ARF/CDKN2A in Mouse, Hamster (Negative), Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
p19ARF | |
Western Blot 1:500, ELISA 1:5000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Ascites | |
RUO | |
Mouse, Hamster (Negative), Human (Negative) | |
Q64364 | |
12578 | |
A synthetic peptide made to a sequence (within residues 1-100) of mouse p19ARF. [UniProt# Q64364] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
19 kDa |
12-A1-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
Ascites with 0.1% Sodium Azide | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
CDKN2A | |
IgG2b | |
Unpurified | |
This antibody is specific for murine p19ARF. It does not recognize human p14Arf nor Golden hamster p13Arf. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title