Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p19ARF/CDKN2A Antibody (5-C3-1) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$207.00 - $487.50
Specifications
Antigen | p19ARF |
---|---|
Clone | 5-C3-1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1 ug/mL, Immunohistochemistry 5 u/gml, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 27177758), Immunohistochemistry-Frozen 5 ug/ml, Flow (Intracellular), CyTOF-ready, Knockout Validated |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB200174SS
![]() |
Novus Biologicals
NB200174SS |
0.025 mL |
Each for $207.00
|
|
|||||
NB200174
![]() |
Novus Biologicals
NB200174 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
p19ARF/CDKN2A Monoclonal specifically detects p19ARF/CDKN2A in Human, Mouse, Golden Syrian Hamster (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready, Knockout Validated.Specifications
p19ARF | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rat | |
ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
CDKN2A | |
IgG2b | |
Protein A purified | |
Mouse. The antibody does not react with human or Golden Syrian hamster. Armenian hamster has not been tested. |
5-C3-1 | |
Western Blot 1:500-1:1000, Flow Cytometry 1 ug/mL, Immunohistochemistry 5 u/gml, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 27177758), Immunohistochemistry-Frozen 5 ug/ml, Flow (Intracellular), CyTOF-ready, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Golden Syrian Hamster (Negative) | |
12578 | |
A synthetic peptide containing amino acids 54-75 of the murine p19ARF. [UniProt# Q64364] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
19 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title