Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p19ARF/CDKN2A Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200106H
Description
p19ARF/CDKN2A Polyclonal specifically detects p19ARF/CDKN2A in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
p16INK4a/CDKN2A | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
Rabbit | |
19 kDa | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
1029 | |
Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HRP | |
PBS with No Preservative | |
CDKN2A | |
Synthetic peptide conjugated to KLH, derived from a sequence within residues 50-150 of mouse p16INK4a. [Swiss-Prot #P51480] | |
Affinity Purified | |
RUO | |
Primary | |
Mouse. Immunogen sequence has 93% homology to rat, 86% homology to porcine and 80% homology to human. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction